(5-Bromo-3-ethoxy-2-fluorophenyl)trimethylsilane ≥97%

Supplier: AOBChem USA

Warning

69198-1G 69198-500MG 69198-5G 69198-25G
77472-764EA 415.09 USD
77472-764 77485-384 77495-964 77505-170
(5-Bromo-3-ethoxy-2-fluorophenyl)trimethylsilane ≥97%
(5-Bromo-3-ethoxy-2-fluorophenyl)trimethylsilane

Precaución: For research use only

Formula: C₁₁H₁₆BrFOSi
Storage Temperature: Ambient
MDL Number: MFCD34763833
CAS Number: 2807450-87-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR