2-Ethoxy-3,4-difluoro-5-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

69148-1G 69148-500MG 69148-5G
77472-756EA 861.08 USD
77472-756 77485-376 77495-956
2-Ethoxy-3,4-difluoro-5-methylbenzaldehyde ≥97%
2-Ethoxy-3,4-difluoro-5-methylbenzaldehyde

Precaución: For research use only

Formula: C₁₀H₁₀F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD34763807
CAS Number: 2807469-66-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR