2-Iodo-5-propylthiophene ≥97%

Supplier: AOBChem USA

Warning

69136-1G 69136-500MG 69136-5G 69136-25G
77472-752EA 284.28 USD
77472-752 77485-372 77495-950 77505-160
2-Iodo-5-propylthiophene ≥97%
2-Iodo-5-propylthiophene

Precaución: For research use only

Formula: C₇H₉IS
Storage Temperature: Ambient
MDL Number: MFCD28668002
CAS Number: 620986-81-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR