2,2-Difluoro-1-(5-propylthiophen-2-yl)ethanone ≥97%

Supplier: AOBChem USA

Warning

69134-1G 69134-500MG 69134-5G 69134-25G
77472-748EA 226.6 USD
77472-748 77485-368 77495-946 77505-156
2,2-Difluoro-1-(5-propylthiophen-2-yl)ethanone ≥97%
2,2-Difluoro-1-(5-propylthiophen-2-yl)ethanone

Precaución: For research use only

Formula: C₉H₁₀F₂OS
MW: 204.237 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763794
CAS Number: 2229251-72-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR