1-(5-ethylthiophen-2-yl)-2,2-difluoroethanone ≥97%

Supplier: AOBChem USA

Warning

69130-1G 69130-500MG 69130-5G 69130-25G
77472-744EA 226.6 USD
77472-744 77485-364 77495-942 77505-152
1-(5-ethylthiophen-2-yl)-2,2-difluoroethanone ≥97%
1-(5-ethylthiophen-2-yl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₈H₈F₂OS
Storage Temperature: Ambient
MDL Number: MFCD28152241
CAS Number: 1692158-90-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR