1-(5-Butylthiophen-2-yl)-2,2-difluoroethanone ≥97%

Supplier: AOBChem USA

Warning

69124-1G 69124-500MG 69124-5G 69124-25G
77472-740EA 82.4 USD
77472-740 77485-360 77495-938 77505-148
1-(5-Butylthiophen-2-yl)-2,2-difluoroethanone ≥97%
1-(5-Butylthiophen-2-yl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₁₀H₁₂F₂OS
MW: 218.263 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763791
CAS Number: 2807455-78-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR