1-(5-Butylthiophen-2-yl)ethanone ≥97%

Supplier: AOBChem USA

Warning

69122-1G 69122-500MG 69122-5G 69122-25G
77472-736EA 255.44 USD
77472-736 77485-356 77495-934 77505-144
1-(5-Butylthiophen-2-yl)ethanone ≥97%
1-(5-Butylthiophen-2-yl)ethanone

Precaución: For research use only

Formula: C₁₀H₁₄OS
MW: 182.283 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30291837
CAS Number: 99172-28-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR