2-Hexyl-5-(methylthio)thiophene ≥97%

Supplier: AOBChem USA

Warning

69120-1G 69120-500MG 69120-5G 69120-25G
77472-732EA 82.4 USD
77472-732 77485-352 77495-930 77505-140
2-Hexyl-5-(methylthio)thiophene ≥97%
2-Hexyl-5-(methylthio)thiophene

Precaución: For research use only

Formula: C₁₁H₁₈S₂
Storage Temperature: Ambient
MDL Number: MFCD34763789
CAS Number: 2807441-58-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR