5-Chloro-2,4'-difluoro-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

69061-1G 69061-500MG 69061-5G
77472-728EA 597.4 USD
77472-728 77485-348 77495-924
5-Chloro-2,4'-difluoro-1,1'-biphenyl ≥95%
5-Chloro-2,4'-difluoro-1,1'-biphenyl

Precaución: For research use only

Formula: C₁₂H₇ClF₂
Storage Temperature: Ambient
MDL Number: MFCD31429347
CAS Number: 2088772-04-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR