(2,3-Difluoro-4-iodophenyl)trimethylsilane ≥97%

Supplier: AOBChem USA

Warning

69040-1G 69040-500MG 69040-5G 69040-25G
77472-724EA 190.55 USD
77472-724 77485-344 77495-920 77505-132
(2,3-Difluoro-4-iodophenyl)trimethylsilane ≥97%
(2,3-Difluoro-4-iodophenyl)trimethylsilane

Precaución: For research use only

Formula: C₉H₁₁F₂ISi
MW: 312.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30178777
CAS Number: 2184302-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR