4-(4-Methylbenzyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

69024-1G 69024-500MG 69024-5G
77472-720EA 688.04 USD
77472-720 77485-340 77495-916
4-(4-Methylbenzyl)benzaldehyde ≥97%
4-(4-Methylbenzyl)benzaldehyde

Precaución: For research use only

Formula: C₁₅H₁₄O
Storage Temperature: Ambient
MDL Number: MFCD34763743
CAS Number: 2248435-26-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR