1-(2,3-Dimethoxypyridin-4-yl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

68941-1G 68941-500MG 68941-5G 68941-25G
77472-716EA 415.09 USD
77472-716 77485-336 77495-910 77505-124
1-(2,3-Dimethoxypyridin-4-yl)-2,2,2-trifluoroethanone ≥95%
1-(2,3-Dimethoxypyridin-4-yl)-2,2,2-trifluoroethanone

Precaución: For research use only

Formula: C₉H₈F₃NO₃
Storage Temperature: Ambient
MDL Number: MFCD34763703
CAS Number: 2918889-48-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR