1-(5-Chloro-2-fluoropyridin-3-yl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

68938-1G 68938-500MG 68938-5G
77472-712EA 517.06 USD
77472-712 77485-332 77495-906
1-(5-Chloro-2-fluoropyridin-3-yl)-2,2,2-trifluoroethanone ≥95%
1-(5-Chloro-2-fluoropyridin-3-yl)-2,2,2-trifluoroethanone

Precaución: For research use only

Formula: C₇H₂ClF₄NO
Storage Temperature: Ambient
MDL Number: MFCD30469228
CAS Number: 2090151-53-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR