1-(3-Bromo-5-chlorophenyl)ethanol ≥97%

Supplier: AOBChem USA

Warning

68864-1G 68864-500MG 68864-5G 68864-25G
77472-708EA 143.17 USD
77472-708 77485-328 77495-900 77505-116
1-(3-Bromo-5-chlorophenyl)ethanol ≥97%
1-(3-Bromo-5-chlorophenyl)ethanol

Precaución: For research use only

Formula: C₈H₈BrClO
Storage Temperature: Ambient
MDL Number: MFCD23140679
CAS Number: 1695104-51-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR