4,5-Dibromobenzo[d][1,3]dioxole ≥97%

Supplier: AOBChem USA

Warning

68846-1G 68846-500MG 68846-5G 68846-25G
77472-704EA 374.92 USD
77472-704 77485-324 77495-896 77505-112
4,5-Dibromobenzo[d][1,3]dioxole ≥97%
4,5-Dibromobenzo[d][1,3]dioxole

Precaución: For research use only

Formula: C₇H₄Br₂O₂
Storage Temperature: Ambient
MDL Number: MFCD02104063
CAS Number: 72744-49-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR