2-Iodo-5-(methylthio)thiophene ≥97%

Supplier: AOBChem USA

Warning

68812-1G 68812-500MG 68812-5G 68812-25G
77472-700EA 134.93 USD
77472-700 77485-320 77495-892 77505-108
2-Iodo-5-(methylthio)thiophene ≥97%
2-Iodo-5-(methylthio)thiophene

Precaución: For research use only

Formula: C₅H₅IS₂
MW: 256.128 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28669239
CAS Number: 86369-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR