4-Bromo-1-butoxy-2-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

68577-1G 68577-500MG 68577-5G 68577-25G
77472-692EA 320.33 USD
77472-692 77485-312 77495-884 77505-100
4-Bromo-1-butoxy-2-methylbenzene ≥95%
4-Bromo-1-butoxy-2-methylbenzene

Precaución: For research use only

Formula: C₁₁H₁₅BrO
MW: 243.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18361006
CAS Number: 287954-84-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR