1-Bromo-2,5-dimethyl-4-propoxybenzene ≥97%

Supplier: AOBChem USA

Warning

68572-1G 68572-500MG 68572-5G 68572-25G
77472-688EA 303.85 USD
77472-688 77485-308 77495-880 77505-096
1-Bromo-2,5-dimethyl-4-propoxybenzene ≥97%
1-Bromo-2,5-dimethyl-4-propoxybenzene

Precaución: For research use only

Formula: C₁₁H₁₅BrO
MW: 243.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763495
CAS Number: 2807471-14-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR