(3-Bromo-2-chloro-5-ethylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

68462-1G 68462-500MG 68462-5G 68462-25G
77472-684EA 365.65 USD
77472-684 77485-304 77495-876 77505-092
(3-Bromo-2-chloro-5-ethylphenyl)(methyl)sulfane ≥97%
(3-Bromo-2-chloro-5-ethylphenyl)(methyl)sulfane

Precaución: For research use only

Formula: C₉H₁₀BrClS
Storage Temperature: Ambient
MDL Number: MFCD34763401
CAS Number: 2707368-81-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR