2-(Benzyloxy)-3-chloro-4-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

68425-1G 68425-500MG 68425-5G 68425-25G
77472-680EA 370.8 USD
77472-680 77485-300 77495-872 77505-090
2-(Benzyloxy)-3-chloro-4-methoxybenzoic acid ≥97%
2-(Benzyloxy)-3-chloro-4-methoxybenzoic acid

Precaución: For research use only

Formula: C₁₅H₁₃ClO₄
MW: 292.714 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763366
CAS Number: 2807463-81-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR