3-Hydroxy-3-phenyl-propionic acid methyl ester ≥97%

Supplier: AOBChem USA

Warning

68360-1G 68360-500MG 68360-5G 68360-25G
77472-676EA 116.39 USD
77472-676 77485-296 77495-868 77505-088
3-Hydroxy-3-phenyl-propionic acid methyl ester ≥97%
3-Hydroxy-3-phenyl-propionic acid methyl ester

Precaución: For research use only

Formula: C₁₀H₁₂O₃
Storage Temperature: Ambient
MDL Number: MFCD19691249
CAS Number: 7497-61-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR