2-Ethoxy-5-fluoro-3-(methylthio)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

68348-1G 68348-500MG 68348-5G 68348-25G
77472-672EA 415.09 USD
77472-672 77485-292 77495-864 77505-084
2-Ethoxy-5-fluoro-3-(methylthio)benzaldehyde ≥95%
2-Ethoxy-5-fluoro-3-(methylthio)benzaldehyde

Precaución: For research use only

Formula: C₁₀H₁₁FO₂S
MW: 214.257 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760946
CAS Number: 2901099-21-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR