3-Fluoro-2-methoxy-4-(methylthio)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

68266-1G 68266-500MG 68266-5G
77472-664EA 479.98 USD
77472-664 77485-284 77495-856
3-Fluoro-2-methoxy-4-(methylthio)benzaldehyde ≥97%
3-Fluoro-2-methoxy-4-(methylthio)benzaldehyde

Precaución: For research use only

Formula: C₉H₉FO₂S
MW: 200.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763279
CAS Number: 2807440-83-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR