1-(6-Bromo-2-chloro-3-methoxyphenyl)-2,2-difluoroethanone ≥95%
Supplier: AOBChem USA
|
Warning
|
68157-1G
68157-500MG
68157-5G
68157-25G
77472-660EA
415.09
USD
77472-660
77485-280
77495-852
77505-078
1-(6-Bromo-2-chloro-3-methoxyphenyl)-2,2-difluoroethanone ≥95%
1-(6-Bromo-2-chloro-3-methoxyphenyl)-2,2-difluoroethanone
Precaución: For research use only
Formula:
C₉H₆BrClF₂O₂ MW: 299.497 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34814791 CAS Number: 2921854-64-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...