1-(3-Bromo-2-ethoxy-6-fluorophenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

68145-1G 68145-500MG 68145-5G 68145-25G
77472-656EA 415.09 USD
77472-656 77485-276 77495-848 77505-074
1-(3-Bromo-2-ethoxy-6-fluorophenyl)-2,2-difluoroethanone ≥95%
1-(3-Bromo-2-ethoxy-6-fluorophenyl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₁₀H₈BrF₃O₂
MW: 297.069 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34814789
CAS Number: 2921686-61-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR