4-Bromo-5-chloro-3-fluoro-2-methoxypyridine ≥97%

Supplier: AOBChem USA

Warning

68104-1G 68104-500MG 68104-5G 68104-25G
77472-652EA 133.9 USD
77472-652 77485-272 77495-842 77505-068
4-Bromo-5-chloro-3-fluoro-2-methoxypyridine ≥97%
4-Bromo-5-chloro-3-fluoro-2-methoxypyridine

Precaución: For research use only

Formula: C₆H₄BrClFNO
MW: 240.457 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763180
CAS Number: 2807454-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR