5-Fluoro-4-isopropoxy-2-(trifluoromethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

68018-1G 68018-500MG 68018-5G
77472-648EA 620.06 USD
77472-648 77485-268 77495-838
5-Fluoro-4-isopropoxy-2-(trifluoromethyl)benzaldehyde ≥97%
5-Fluoro-4-isopropoxy-2-(trifluoromethyl)benzaldehyde

Precaución: For research use only

Formula: C₁₁H₁₀F₄O₂
MW: 250.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763120
CAS Number: 2918941-51-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR