3-Fluoro-5-methoxy-4-(methylthio)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

67998-1G 67998-500MG 67998-5G 67998-25G
77472-644EA 415.09 USD
77472-644 77485-264 77495-834 77505-064
3-Fluoro-5-methoxy-4-(methylthio)benzaldehyde ≥97%
3-Fluoro-5-methoxy-4-(methylthio)benzaldehyde

Precaución: For research use only

Formula: C₉H₉FO₂S
MW: 200.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18904762
CAS Number: 1379315-46-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR