5-Bromo-4-(m-tolyl)benzo[d][1,3]dioxole ≥97%

Supplier: AOBChem USA

Warning

67974-1G 67974-500MG 67974-5G 67974-25G
77472-640EA 415.09 USD
77472-640 77485-260 77495-830 77505-062
5-Bromo-4-(m-tolyl)benzo[d][1,3]dioxole ≥97%
5-Bromo-4-(m-tolyl)benzo[d][1,3]dioxole

Precaución: For research use only

Formula: C₁₄H₁₁BrO₂
Storage Temperature: Ambient
MDL Number: MFCD34763089
CAS Number: 2901102-49-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR