1-(3-Chloro-5-fluorophenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

67896-1G 67896-500MG 67896-5G 67896-25G
77472-636EA 264.71 USD
77472-636 77485-256 77495-826 77505-058
1-(3-Chloro-5-fluorophenyl)-2,2-difluoroethanone ≥95%
1-(3-Chloro-5-fluorophenyl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₈H₄ClF₃O
Storage Temperature: Ambient
MDL Number: MFCD20920396
CAS Number: 1352208-84-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR