5’-Bromo-2’-fluoro-3’-(trifluoromethyl)acetophenone ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 1-(5-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)ethanone

67812-1G 67812-500MG 67812-5G
77472-628EA 586.07 USD
77472-628 77485-248 77495-818
5’-Bromo-2’-fluoro-3’-(trifluoromethyl)acetophenone ≥95%
5’-Bromo-2’-fluoro-3’-(trifluoromethyl)acetophenone

Precaución: For research use only

Formula: C₉H₅BrF₄O
MW: 285.033 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32899472
CAS Number: 2092468-17-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR