2-(Difluoromethoxy)-5-fluorobenzoic acid ≥95%

Supplier: AOBChem USA

Warning

67737-1G 67737-500MG 67737-5G 67737-25G
77472-624EA 147.29 USD
77472-624 77485-244 77495-814 77505-050
2-(Difluoromethoxy)-5-fluorobenzoic acid ≥95%
2-(Difluoromethoxy)-5-fluorobenzoic acid

Precaución: For research use only

Formula: C₈H₅F₃O₃
Storage Temperature: Ambient
MDL Number: MFCD09832364
CAS Number: 1017779-44-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR