4-Fluoro-5-(trifluoromethyl)-[1,1'-biphenyl]-3-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

67630-1G 67630-500MG 67630-5G 67630-25G
77472-616EA 415.09 USD
77472-616 77485-236 77495-806 77505-042
4-Fluoro-5-(trifluoromethyl)-[1,1'-biphenyl]-3-carbaldehyde ≥97%
4-Fluoro-5-(trifluoromethyl)-[1,1'-biphenyl]-3-carbaldehyde

Precaución: For research use only

Formula: C₁₄H₈F₄O
MW: 268.206 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762918
CAS Number: 2901066-77-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR