3-Bromo-2-chloro-6-fluoro-5-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

67551-1G 67551-500MG 67551-5G 67551-25G
77472-608EA 226.6 USD
77472-608 77485-228 77495-798 77505-036
3-Bromo-2-chloro-6-fluoro-5-methylbenzaldehyde ≥97%
3-Bromo-2-chloro-6-fluoro-5-methylbenzaldehyde

Precaución: For research use only

Formula: C₈H₅BrClFO
Storage Temperature: Ambient
MDL Number: MFCD34762876
CAS Number: 2807455-69-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR