2-(3-Bromo-5-fluorophenyl)-2-methyl-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

67529-1G 67529-5G 67529-25G
77472-604EA 60.77 USD
77472-604 77495-794 77505-032
2-(3-Bromo-5-fluorophenyl)-2-methyl-1,3-dioxolane ≥97%
2-(3-Bromo-5-fluorophenyl)-2-methyl-1,3-dioxolane

Precaución: For research use only

Formula: C₁₀H₁₀BrFO₂
MW: 261.088 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762864
CAS Number: 2901098-61-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR