1-(3-Bromo-2-fluoro-5-methylphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

67495-1G 67495-500MG 67495-5G 67495-25G
77472-600EA 370.8 USD
77472-600 77485-224 77495-790 77505-028
1-(3-Bromo-2-fluoro-5-methylphenyl)ethanone ≥95%
1-(3-Bromo-2-fluoro-5-methylphenyl)ethanone

Precaución: For research use only

Formula: C₉H₈BrFO
MW: 231.062 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28130740
CAS Number: 1514822-22-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR