1-(2,3-Difluoro-6-methoxyphenyl)-2,2-Difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

67444-1G 67444-500MG 67444-5G 67444-25G
77472-596EA 415.09 USD
77472-596 77485-220 77495-786 77505-024
1-(2,3-Difluoro-6-methoxyphenyl)-2,2-Difluoroethanone ≥95%
1-(2,3-Difluoro-6-methoxyphenyl)-2,2-Difluoroethanone

Precaución: For research use only

Formula: C₉H₆F₄O₂
MW: 222.136 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34814680
CAS Number: 2921840-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR