1-(4'-Bromo-[1,1'-biphenyl]-4-yl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

67227-1G 67227-500MG 67227-5G 67227-25G
77472-588EA 382.13 USD
77472-588 77485-212 77495-778 77505-018
1-(4'-Bromo-[1,1'-biphenyl]-4-yl)-2,2,2-trifluoroethanone ≥95%
1-(4'-Bromo-[1,1'-biphenyl]-4-yl)-2,2,2-trifluoroethanone

Precaución: For research use only

Formula: C₁₄H₈BrF₃O
Storage Temperature: Ambient
MDL Number: MFCD34814652
CAS Number: 206758-26-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR