2,2,2-Trifluoro-1-(2-fluoro-3-iodophenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

67172-1G 67172-500MG 67172-5G 67172-25G
77472-584EA 415.09 USD
77472-584 77485-208 77495-774 77505-016
2,2,2-Trifluoro-1-(2-fluoro-3-iodophenyl)ethanone ≥95%
2,2,2-Trifluoro-1-(2-fluoro-3-iodophenyl)ethanone

Precaución: For research use only

Formula: C₈H₃F₄IO
MW: 318.007 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28130778
CAS Number: 1823324-67-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR