1-(3,5-Di-tert-butylphenyl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

66921-1G 66921-500MG 66921-5G
77472-576EA 442.9 USD
77472-576 77485-200 77495-766
1-(3,5-Di-tert-butylphenyl)-2,2,2-trifluoroethanone ≥95%
1-(3,5-Di-tert-butylphenyl)-2,2,2-trifluoroethanone

Precaución: For research use only

Formula: C₁₆H₂₁F₃O
Storage Temperature: Ambient
MDL Number: MFCD11556173
CAS Number: 1128165-27-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR