2,2,2-Trifluoro-1-(2-methoxy-4-(trifluoromethyl)phenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

66803-1G 66803-500MG 66803-5G
77472-568EA 746.75 USD
77472-568 77485-192 77495-758
2,2,2-Trifluoro-1-(2-methoxy-4-(trifluoromethyl)phenyl)ethanone ≥95%
2,2,2-Trifluoro-1-(2-methoxy-4-(trifluoromethyl)phenyl)ethanone

Precaución: For research use only

Formula: C₁₀H₆F₆O₂
MW: 272.144 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28131784
CAS Number: 1551495-63-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR