2,2-Difluoro-1-(2-fluoro-6-methoxy-3-methylphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

66777-1G 66777-500MG 66777-5G 66777-25G
77472-564EA 415.09 USD
77472-564 77485-188 77495-754 77505-012
2,2-Difluoro-1-(2-fluoro-6-methoxy-3-methylphenyl)ethanone ≥95%
2,2-Difluoro-1-(2-fluoro-6-methoxy-3-methylphenyl)ethanone

Precaución: For research use only

Formula: C₁₀H₉F₃O₂
MW: 218.172 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34814559
CAS Number: 2921694-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR