2-(3-Chloro-4-fluoro-5-methylphenyl)propan-2-ol ≥97%

Supplier: AOBChem USA

Warning

66752-1G 66752-500MG 66752-5G
77472-560EA 546.93 USD
77472-560 77485-184 77495-750
2-(3-Chloro-4-fluoro-5-methylphenyl)propan-2-ol ≥97%
2-(3-Chloro-4-fluoro-5-methylphenyl)propan-2-ol

Precaución: For research use only

Formula: C₁₀H₁₂ClFO
MW: 202.653 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28526648
CAS Number: 1783525-64-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR