(4-(Benzyloxy)-2-chlorophenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

66725-1G 66725-500MG 66725-5G 66725-25G
77472-556EA 415.09 USD
77472-556 77485-180 77495-746 77505-006
(4-(Benzyloxy)-2-chlorophenyl)(methyl)sulfane ≥95%
(4-(Benzyloxy)-2-chlorophenyl)(methyl)sulfane

Precaución: For research use only

Formula: C₁₄H₁₃ClOS
MW: 264.77 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762749
CAS Number: 2918828-73-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR