1-Bromo-2-(methoxymethoxy)-3,5-bis(trifluoromethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

66629-1G 66629-500MG 66629-5G
77472-552EA 515 USD
77472-552 77485-176 77495-742
1-Bromo-2-(methoxymethoxy)-3,5-bis(trifluoromethyl)benzene ≥95%
1-Bromo-2-(methoxymethoxy)-3,5-bis(trifluoromethyl)benzene

Precaución: For research use only

Formula: C₁₀H₇BrF₆O₂
MW: 353.056 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762699
CAS Number: 2807471-43-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR