6-Isobutoxynicotinaldehyde ≥97%

Supplier: AOBChem USA

Warning

66465-1G 66465-500MG 66465-5G
77472-548EA 914.64 USD
77472-548 77485-172 77495-738
6-Isobutoxynicotinaldehyde ≥97%
6-Isobutoxynicotinaldehyde

Precaución: For research use only

Formula: C₁₀H₁₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD23952666
CAS Number: 1427081-01-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR