2,4,5-Tribromo-1-butyl-1H-imidazole ≥97%

Supplier: AOBChem USA

Warning

66457-1G 66457-500MG 66457-5G 66457-25G
77472-544EA 197.76 USD
77472-544 77485-168 77495-732 77504-996
2,4,5-Tribromo-1-butyl-1H-imidazole ≥97%
2,4,5-Tribromo-1-butyl-1H-imidazole

Precaución: For research use only

Formula: C₇H₉Br₃N₂
MW: 360.872 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30304790
CAS Number: 2302-24-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR