1-(3,4-Dichloro-2-fluorophenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

66379-1G 66379-500MG 66379-5G
77472-540EA 438.78 USD
77472-540 77485-164 77495-728
1-(3,4-Dichloro-2-fluorophenyl)ethanone ≥95%
1-(3,4-Dichloro-2-fluorophenyl)ethanone

Precaución: For research use only

Formula: C₈H₅Cl₂FO
Storage Temperature: Ambient
MDL Number: MFCD28785938
CAS Number: 1806354-87-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR