1-(6-Bromo-2-chloro-3-methylphenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

66319-1G 66319-500MG 66319-5G 66319-25G
77472-536EA 415.09 USD
77472-536 77485-160 77495-724 77504-992
1-(6-Bromo-2-chloro-3-methylphenyl)-2,2-difluoroethanone ≥95%
1-(6-Bromo-2-chloro-3-methylphenyl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₉H₆BrClF₂O
MW: 283.497 g/mol
Storage Temperature: Ambient
CAS Number: 3002527-79-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR