2-Bromo-4-chloro-1-(difluoromethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

66279-1G 66279-500MG 66279-5G
77472-532EA 849.75 USD
77472-532 77485-156 77495-720
2-Bromo-4-chloro-1-(difluoromethyl)benzene ≥95%
2-Bromo-4-chloro-1-(difluoromethyl)benzene

Precaución: For research use only

Formula: C₇H₄BrClF₂
Storage Temperature: Ambient
MDL Number: MFCD18391081
CAS Number: 1261476-50-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR